missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PXDN Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144080
This item is not returnable.
View return policy
Description
Positive Control - WB: human Hela whole cell, human U251 whole cell, rat heart tissue, mouse heart tissue. IHC: human pancrease cancer tissue, human renal cancer tissue.
Drosophila peroxidasin is an extracellular matrix-associated peroxidase (Horikoshi et al., 1999 [PubMed 10441517]). It is expressed exclusively in hemocytes derived from head mesoderm at a very early stage of differentiation. Peroxidasin exists as a homotrimer with a unique hybrid structure that combines an enzymatically functional peroxidase domain with motifs that are typically found in extracellular matrix-associated proteins. It is a secreted protein that contains a secretory recognition sequence at its N terminus. Peroxidasin catalyzes hydrogen peroxide-driven radioiodination, oxidations, and the formation of dityrosine in vitro. It is also thought to function in extracellular matrix consolidation, phagocytosis, and defense.
Specifications
| PXDN | |
| Polyclonal | |
| Unconjugated | |
| PXDN | |
| 2310075M15Rik; C85409; COPOA; D2S448; D2S448E; E330004E07; Kiaa0230; Melanoma-associated antigen MG50; MG50; mKIAA0230; p53-responsive gene 2 protein; PE responsive protein c64; Perc64; peroxidasin; peroxidasin homolog; peroxidasin homolog (Drosophila); PRG2; PXDN; PXN; Vascular peroxidase 1; VPO; VPO1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 554172, 69675, 7837 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q3UQ28, Q92626 | |
| PXDN | |
| Human PXDN between 900-1064. | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction