missing translation for 'onlineSavingsMsg'
Learn More

(-)-PX20606 trans isomer, MedChemExpress

Product Code. p-200101406
Encompass_Preferred
Click to view available options
Quantity:
1 mg
2 mg
Unit Size:
1mg
2mg
This item is not returnable. View return policy

Product Code. 16490649

Brand: Medchem Express HY100443B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (-)-PX20606 trans isomer is a FXR agonist with EC50s of 18 and 29 nM for FXR in FRET and M1H assay, respectively.

(-)-PX20606 trans isomer is a FXR agonist with EC50s of 18 and 29 nM for FXR in FRET and M1H assay, respectively.

Specifications

Chemical Name or Material (-)-PX20606 trans isomer
CAS 1268244-88-7
Purity Grade Notes Research
Molecular Formula C29H22Cl3NO4
Quantity 1 mg
Synonym (-)-PX-102 trans isomer (-)-PX-104
Solubility Information Ethanol : ≥ 50 mg/mL (90.11 mM)
SMILES ClC1=C(C2=NOC(C3CC3)=C2COC4=CC=C([C@@H]5C[C@H]5C6=CC=C(C(O)=O)C=C6)C(Cl)=C4)C(Cl)=CC=C1.[(-)].[Rotation]
Molecular Weight (g/mol) 554.85
Formula Weight 554.85
Grade Research
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.