missing translation for 'onlineSavingsMsg'
Learn More

(-)-PX20606 trans isomer, MedChemExpress

Product Code. 16490649
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
2 mg
Unit Size:
1mg
2mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490649 1 mg 1mg
16470016 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16490649 Supplier Medchem Express Supplier No. HY100443B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (-)-PX20606 trans isomer is a FXR agonist with EC50s of 18 and 29 nM for FXR in FRET and M1H assay, respectively.

(-)-PX20606 trans isomer is a FXR agonist with EC50s of 18 and 29 nM for FXR in FRET and M1H assay, respectively.

Specifications

Chemical Name or Material (-)-PX20606 trans isomer
CAS 1268244-88-7
Purity Grade Notes Research
Molecular Formula C29H22Cl3NO4
Quantity 1 mg
Synonym (-)-PX-102 trans isomer (-)-PX-104
Solubility Information Ethanol : ≥ 50 mg/mL (90.11 mM)
SMILES ClC1=C(C2=NOC(C3CC3)=C2COC4=CC=C([C@@H]5C[C@H]5C6=CC=C(C(O)=O)C=C6)C(Cl)=C4)C(Cl)=CC=C1.[(-)].[Rotation]
Molecular Weight (g/mol) 554.85
Formula Weight 554.85
Grade Research
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.