missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PX 12, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Thioredoxin-1 (Trx-1) inhibitor; irreversibly inactivates Trx-1 as a substrate for reduction by thioredoxin reductase (TR); competitively inhibits the reduction of Trx-1 by TR (Ki = 30.8 μM). Attenuates expression of HIF-1α, VEGF and iNOS (IC50 values are 7.2, 10.4 and 18.1 μM respectively). Exhibits antitumor activity; directly inhibits tubulin in vitro and decreases tumor microvessel density in vivo.
Specifications
Quantity | 10 mg |
Formula Weight | 188.31 |
Percent Purity | >99% |
Chemical Name or Material | PX 12 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'