missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PWP1 Antibody (S01-7B5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19752-50ul
This item is not returnable.
View return policy
Description
PWP1 Monoclonal antibody specifically detects PWP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), RNA Inhibition, Sandwich ELISA
Specifications
| PWP1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry 1:10-1:500, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, RNA Inhibition, Sandwich ELISA 1:100-1:2000 | |
| IEF-SSP-9502, Keratinocyte protein IEF SSP 9502, nuclear phosphoprotein similar to S. cerevisiae PWP1, periodic tryptophan protein 1 homolog, PWP1 homolog (S. cerevisiae) | |
| A synthetic peptide of human PWP1 (Uniprot # Q13610) | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| S01-7B5 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 11137 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction