missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PVR (CD155) Mouse anti-Human, Oligo, Clone: TX24, BD™ AbSeq ™
Mouse Monoclonal Antibody
Brand: BD Biosciences 940102
This item is not returnable.
View return policy
Description
PVR (CD155) Monoclonal antibody specifically detects PVR (CD155) in Human samples. It is validated for Single Cell 3' Sequencing
Specifications
| PVR (CD155) | |
| Monoclonal | |
| Oligo | |
| Poliovirus receptor; HVED; Nectin-like protein 5; NECL5; NECL-5; PVS; TAGE4 | |
| Human PVR Transfected Cell Line | |
| RUO | |
| Human | |
| IgG2a |
| Single Cell 3' Sequencing | |
| Clone TX24 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction