Learn More
Masterflex™ Barbed Y-Connector, PVDF
Combine these simple, economical fittings with flexible tubing for use in low-pressure applications (generally less than 30 psi). Cole-Parmer™ PVDF Barbed Y-Connectors are designed for quick and easy assembly without tools.
Brand:  Masterflex™ 30703-90
Product Code. 11806643
Quantity Price
1 £32.62 / Pack of 10
10 £29.96 / Pack of 10
In Stock
Edge
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Temperature range: –62 to 129°C
  • Excellent Chemical Resistance
  • Sterilize by ethylene oxide or autoclave