missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ PVDF Sanitary Tee

Product Code. 11734979
Change view
Click to view available options
Clamp Size: 0.75in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11734979 Clamp Size: 0.75in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11734979 Supplier Masterflex™ Supplier No. 3180961

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Autoclavable Autoclavable
Material PVDF
Min. Operating Temperature (Metric) -40°C
Packaging Quantity 1 Pack
Max. Operating Temperature (Metric) 138°C
Product Type Sanitary Fitting
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.