Learn More
Faster™ PVC Shutter for Front Aperture of FlowFAST H Safety Cabinet
Product Code. 15503336
Click to view available options
For Use With (Equipment):
FlowFAST H 09
FlowFAST H 12
FlowFAST H 15
FlowFAST H 18
Product Code. 15503336
Brand: Faster™ FX0000224580
To allow closure of the opening of the FlowFAST cabinet.
  • PVC shutter
  • Manual pull down shutter
Specifications
Certifications/Compliance EN 61010-2001
For Use With (Equipment) FlowFAST H 18