Learn More
Dostmann Electronic™ PVC Precision Probes
Product Code. 11854402
Click to view available options
Sensor Types:
Class 1/10
Class 1/3
Unit Size:
Each
Product Code. 11854402
Brand: Dostmann Electronic™ 60001074
Only null left
Add to basket
Add to basket
Rich, robust accessory for hand-held meter Series P700 that is ideal for use in many laboratory, scientific, and research environments.
  • Temperature Sensor Pt100 6000-1074
  • Pt100, Cl. 1/10 DIN -200...+450 °C 300 x 3,0 mm
  • Immer­sion probe for meas­uring in liquid and powderd mater­ials
  • Sensor Class 1/​10, Pt100(ceramic), 4-wire, DIN IEC 751,
  • tube
  • V2A or inconel, mineral-insu­lated, with handle, thermal stabil­ized
Specifications
Type Probe, Precision
Material PVC
Sensor Types Class 1/10
Accuracy ± 0.03°C
Temperature Range (Metric) -200°C to +450°C
Length (Metric) 300 mm