missing translation for 'onlineSavingsMsg'
Learn More

Brady™ PVC ISO First Aid Signs

Product Code. 15103289
Click to view available options
Style:
Bi-directional
Panoramic
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15103289

Brand: Brady™ 814613

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Certifications/Compliance ISO 7010
Alternate Part Number PIC E003-203x203-BIPVC-CRD/1
Material PVC
Quantity 1 Ea.
Style Bi-directional
For Use With (Application) Indoor & Outdoor
Height (Metric) 203 mm
Thickness 1.591 mm
Width (Metric) 203.00 mm
Temperature Range (Metric) -20 °C-70 °C
Shape Square
Languages Global
Color White on Green
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.