missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ PVC Hazardous Label Roll

Mark hazardous material warning messages on containers, instruments and equipment when hazardous materials are in use. Each Allsigns International™ PVC Hazardous Label Roll features a distinct warning message on a roll of self-adhesive vinyl “diamond” labels.

Brand:  Allsigns International™ DA9

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12395768

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Sold in single rolls
  • Self-adhesive vinyl
  • Perforated for ease of removal
Specifications

Specifications

Material: Perforated vinyl
Toxic 6
Self-adhesive
Chemicals
102mm
102 mm
Vinyl
Videos
Documentation