missing translation for 'onlineSavingsMsg'
Learn More

HUBER PVC Hoses

Pressureless PVC hose

Brand:  HUBER 6071

 View more versions of this product

Product Code. 10759372

  • £39.91 / Each
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • PVC
  • Min. temp.: -20°C
  • Max. temp.: 60°C
  • Specifications

    Specifications

    PVC
    -20°C
    8 mm
    60°C
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers