missing translation for 'onlineSavingsMsg'
Learn More

Putrescine dihydrochloride, MP Biomedicals™

Product Code. 11493130 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
Unit Size:
100g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493130 1 g 1g
11403140 25 g 25g
11413140 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493130 Supplier MP Biomedicals Supplier No. 0210045001
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Putrescine is a biogenic polyamine and precursor of spermidine. It is formed via decarboxylation of ornithine or by decarboxylation of arginine followed by hydrolysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-93-7
Molecular Formula C4H14Cl2N2
Molecular Weight (g/mol) 161.07
MDL Number MFCD00012526
InChI Key XXWCODXIQWIHQN-UHFFFAOYSA-N
Synonym 1,4-diaminobutane dihydrochloride, putrescine dihydrochloride, tetramethylenediamine dihydrochloride, putrescine hydrochloride, 1,4-butanediamine dihydrochloride, unii-x45sur7rhy, 1,4-butanediamine, dihydrochloride, 1,4-butanediamine, hydrochloride, 1,4-diaminobutane, dihydrochloride, butane-1,4-diamine dihydrochloride
PubChem CID 9532
IUPAC Name butane-1,4-diamine;dihydrochloride
SMILES C(CCN)CN.Cl.Cl

Specifications

Color White
Quantity 1 g
Formula Weight 161.08
Physical Form Crystalline Powder
Chemical Name or Material Putrescine dihydrochloride
compliance-icons
Product Identifier
  • Putrescine Dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: tetramethylenediammonium dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.