missing translation for 'onlineSavingsMsg'
Learn More

Push/Pull Spring Balance

Product Code. 12727056
Encompass_Preferred
Change view
Click to view available options
Capacity:
1000 g
2000 g
5000 g
Unit Size:
Each
Product Code. Capacity unitSize
12727056 2000 g Each
12717056 1000 g -
12747056 5000 g -
3 options
This item is not returnable. View return policy

Product Code. 12727056

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Red
For Use With (Application) For Weighing and for Measuring Force of Pushing or
Capacity 2000 g
Type Push/Pull Spring Balance
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.