missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Push Broom

Product Code. 12755725 Shop All Micronova Products
Change view
Click to view available options
Color:
White
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
12755725 White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12755725 Supplier Micronova™ Supplier No. BROOM24

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The Push Broom combines the features of fine, medium and heavy floor sweeps into one convenient, all-purpose floor sweep.

  • Non-absorbent hygienic plastic block
  • Synthetic bristles with angled pitch for fast, effective sweeping on all floor types
  • Short heavy bristles in front help sweep heavy debris
  • Long, fine/medium bristles in back sweep dust and particles
  • Can be used with sweeping compounds
  • For use with STA series plastic handles

Specifications

Product Type Synthetic Bristle Push Broom Head
Color White
Length (English) 24 in. Broom Head
For Use With (Application) For use with STA series plastic handles.
Certifications/Compliance Base Resins to Produce Blocks and Bristles are made of FDS approved materials. BPA Free.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.