missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PUS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17248-1-AP-150UL
This item is not returnable.
View return policy
Description
Formation of pseudouridine at position 39 in the anticodon stem and loop of transfer RNAs.Specifications
| PUS3 | |
| Polyclonal | |
| Unconjugated | |
| PUS3 | |
| 2610020J05Rik, FKSG32, pseudouridylate synthase 3, PUS3, tRNA pseudouridine synthase 3, tRNA uridine isomerase 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 315554, 67049, 83480 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BZE2, Q9JI38 | |
| Pus3 | |
| PUS3 Fusion Protein Ag9983 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction