missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PUS3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566634
This item is not returnable.
View return policy
Description
Immunogen sequence: DLRSQFPRGRD SEDFNVKEEA NAAAEEIRYT HILNRVLPPD IRILAWAPVE PSFSARFSCL ERTYRYFFPR ADLDIVTMDY AAQKYVGTH Highest antigen sequence identity to the following orthologs - mouse 86%, rat 82%.
Formation of pseudouridine at position 39 in the anticodon stem and loop of transfer RNAs.
Specifications
| PUS3 | |
| Polyclonal | |
| Unconjugated | |
| PUS3 | |
| 2610020J05Rik; 5730412F04Rik; FKSG32; pseudouridine synthase 3; pseudouridylate synthase 3; Pus3; RGD1310757; tRNA pseudouridine synthase 3; tRNA pseudouridine(38/39) synthase; tRNA pseudouridylate synthase 3; tRNA-uridine isomerase 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 83480 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BZE2 | |
| PUS3 | |
| Recombinant Human PUS3. Recombinant protein control fragment (Product #RP-105288). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction