missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PUS10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550738
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total PUS10 protein.
Pseudouridylate synthases catalyze pseudouridination of structural RNAs, including transfer, ribosomal, and splicing RNAs. PUS10 catalyzes the formation of the universal psi55 in the GC loop of transfer RNAs (Probable). Modulator of TRAIL-induced cell death via activation of procaspase 8 and BID cleavage. Required for the progression of the apoptotic signal through intrinsic mitochondrial cell death.
Specifications
| PUS10 | |
| Polyclonal | |
| Unconjugated | |
| Pus10 | |
| 2810013G11Rik; 4933435A13Rik; AU014648; C77560; CCDC139; coiled-coil domain containing 139; coiled-coil domain-containing protein 139; DOBI; pseudouridine synthase 10; pseudouridylate synthase 10; psi55 synthase; Pus10; Putative tRNA pseudouridine synthase Pus10; RGD1306402; tRNA pseudouridine 55 synthase; tRNA pseudouridylate synthase; tRNA-uridine isomerase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 150962, 74467 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2.8 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q3MIT2, Q9D3U0 | |
| Pus10 | |
| Synthetic peptide corresponding to a region derived from internal residues of human pseudouridylate synthase 10. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction