missing translation for 'onlineSavingsMsg'
Learn More

PUS10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-35626-20ul

 View more versions of this product

Product Code. 30229910

  • £145.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PUS10 Polyclonal antibody specifically detects PUS10 in Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PUS10
Polyclonal
Western Blot 1:500 - 1:2000, ELISA
CCDC139, coiled-coil domain containing 139, Coiled-coil domain-containing protein 139, DOBIMGC126729, EC 5.4.99, EC 5.4.99.-, FLJ32312, MGC126755, pseudouridine synthase 10, pseudouridylate synthase 10, Psi55 synthase, PUS1, putative tRNA pseudouridine synthase Pus10, tRNA pseudouridine 55 synthase, tRNA pseudouridylate synthase, tRNA-uridine isomerase
Recombinant fusion protein containing a sequence corresponding to amino acids 290-529 of human PUS10 (NP_653310.2).,, Sequence:, VAGRYNKYSRNLPQTPWIIDGERKLESSVEELISDHLLAVFKAESFNFSSSGREDVDVRTLGNGRPFAIELVNPHRVHFTSQEIKELQQKINNSSNKIQVRDLQLVTREAIGHMKEGEEEKTKTYSALIWTNKAIQKKDIEFLNDIKDLKIDQKTPLRVLHRRPLAVRARVIHFMETQYVDEHHFRLHLKTQAGTYIKEFVHGDFGRTKPNIGSLMNVTADILELDVESVDVDWPPALDD
20 μL
Primary
Mouse, Rat
Purified
ELISA, Western Blot
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
150962
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.