missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PUS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11512-1-AP-20UL
This item is not returnable.
View return policy
Description
The PRKDC gene encodes the catalytic subunit of a nuclear DNA-dependent serine/threonine protein kinase (DNA-PK). The second component is the autoimmune antigen Ku, which is encoded by the G22P1 gene on chromosome 22q. On its own, the catalytic subunit of DNA-PK is inactive and relies on the G22P1 component to direct it to the DNA and trigger its kinase activity; PRKDC must be bound to DNA to express its catalytic properties.Specifications
| PUS1 | |
| Polyclonal | |
| Unconjugated | |
| PUS1 | |
| MLASA, pseudouridylate synthase 1, PUS1, tRNA uridine isomerase I | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 304567, 56361, 80324 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q4KM92, Q9WU56, Q9Y606 | |
| Pus1 | |
| PUS1 Fusion Protein Ag2071 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction