missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PUS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568838
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Mouse: 100%; Rat: 100%.
The PRKDC gene encodes the catalytic subunit of a nuclear DNA-dependent serine/threonine protein kinase (DNA-PK). The second component is the autoimmune antigen Ku, which is encoded by the G22P1 gene on chromosome 22q. On its own, the catalytic subunit of DNA-PK is inactive and relies on the G22P1 component to direct it to the DNA and trigger its kinase activity; PRKDC must be bound to DNA to express its catalytic properties.
Specifications
| PUS1 | |
| Polyclonal | |
| Unconjugated | |
| PUS1 | |
| A730013B20Rik; mitochondrial tRNA pseudouridine synthase A; MLASA1; MNCb-0873; MPUS1; mPus1p; PP8985; pseudouridine synthase 1; pseudouridylate synthase 1; Pus1; pus1 {ECO:0000312; RGD:1311871}; tRNA pseudouridine synthase A; tRNA pseudouridine synthase A, mitochondrial; tRNA pseudouridine(38-40) synthase; tRNA pseudouridylate synthase I; tRNA uridine isomerase I; tRNA-uridine isomerase I | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 56361 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9WU56 | |
| PUS1 | |
| synthetic peptide corresponding to a region of mouse PUS1 (aa 51-100). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido