missing translation for 'onlineSavingsMsg'
Learn More

Purpurin (85%), TRC

Product Code. 30136196
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136196 5 g 5g
30132930 10 g 10g
17984045 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136196 Supplier Toronto Research Chemicals Supplier No. TRCP8403305G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-54-9
Molecular Formula C14 H8 O5
Molecular Weight (g/mol) 256.21
Synonym Anthraquinone, 1,2,4-trihydroxy- (7CI,8CI), Purpurin (6CI), 1,2,4-Trihydroxy-9,10-anthracenedione, 1,2,4-Trihydroxy-9,10-anthraquinone, 1,2,4-Trihydroxy-9,10-dihydroanthracene-9,10-dione, 1,2,4-Trihydroxyanthraquinone, C.I. 58205, C.I. 75410, Hydroxylizaric acid, NSC 10447, Purpurin (dye), Purpurine, Smoke Brown G, Verantin
IUPAC Name 1,2,4-trihydroxyanthracene-9,10-dione
SMILES Oc1cc(O)c2C(=O)c3ccccc3C(=O)c2c1O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C14H8O5/c15-8-5-9(16)14(19)11-10(8)12(17)6-3-1-2-4-7(6)13(11)18/h1-5,15-16,19H
Formula Weight 256.0372
Chemical Name or Material Purpurin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.