missing translation for 'onlineSavingsMsg'
Learn More

Purpureaside C, MedChemExpress

Product Code. 16461738
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16461738 5 mg 5mg
16419308 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16461738 Supplier Medchem Express Supplier No. HYN4148

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Purpureaside C is a phenolic glycoside and has significant proinflammatory action.

Purpureaside C is a phenolic glycoside and has significant proinflammatory action.

Specifications

Chemical Name or Material Purpureaside C
CAS 108648-07-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C35H46O20
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (158.89 mM; Need ultrasonic)
SMILES OC1=C(O)C=CC(/C=C/C(O[C@H]2[C@@H]([C@H]([C@H](OCCC3=CC(O)=C(O)C=C3)O[C@@H]2CO[C@@H]4O[C@@H]([C@H](O)[C@H](O)[C@H]4O)CO)O)O[C@@]5([H])[C@@H]([C@@H]([C@@H](O)[C@H](C)O5)O)O)=O)=C1
Molecular Weight (g/mol) 786.73
Formula Weight 786.73
Percent Purity 94.42%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.