missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Puromycin Dihydrochloride,, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mg
25 mg
100 mg
250 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
250mg
25mg
500mg
Specifications
Specifications
| Type | Puromycin Dihydrochloride |
| Physical Form | Powder |
| Extinction Coefficient | 20.3 (275nm, 0.1N NaOH), 19.5 (268nm, 0.1 NHCl) (Lit.) |
| Color | White |
| Quantity | 1 g |
| IUPAC Name | (2S)-2-amino-N-[(2S,3S,4S,5R)-5-[6-(dimethylamino)purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide;dihydrochloride |
| Formula Weight | 544.4 |
| Chemical Name or Material | Puromycin Dihydrochloride, from Streptomyces albo-niger |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction