missing translation for 'onlineSavingsMsg'
Learn More

Melford Puromycin Dihydrochloride

Product Code. 30018692
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018692 500 mg 500mg
30018691 100 mg 100mg
30018690 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018692 Supplier Melford Supplier No. P330200.5
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puromycin Dihydrochloride is a potent antibiotic.

Puromycin Dihydrochloride is a potent antibiotic. It is an amino nucleoside antibiotic that mimics the 3'-end of aminoacyl-tRNA. It enters the ribosomal A site during translation, causing premature chain termination and inhibiting protein synthesis. Puromycin is incorporated into growing polypeptide chains, leading to the release of incomplete and nonfunctional peptides.

Widely used as a tool in cell biology to study protein synthesis and investigate translation processes. Its ability to terminate protein synthesis at the ribosome makes it valuable for various applications, including pulse-chase experiments and the analysis of nascent peptides. Puromycin resistance genes are often used as selectable markers in genetic engineering. Cells expressing the resistance gene can survive and proliferate in the presence of Puromycin, allowing for the selection of transfected or transduced cells.

Puromycin Dihydrochloride is a valuable tool for investigating protein synthesis, studying translation mechanisms, and selecting cells with specific genetic modifications.

Specifications

Chemical Name or Material Puromycin
CAS 58-58-2
Quantity 500 mg
Molecular Formula C22H29N7O5-2 HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.