Learn More
Melford Puromycin Dihydrochloride
Description
Puromycin Dihydrochloride is a potent antibiotic.
Widely used as a tool in cell biology to study protein synthesis and investigate translation processes. Its ability to terminate protein synthesis at the ribosome makes it valuable for various applications, including pulse-chase experiments and the analysis of nascent peptides. Puromycin resistance genes are often used as selectable markers in genetic engineering. Cells expressing the resistance gene can survive and proliferate in the presence of Puromycin, allowing for the selection of transfected or transduced cells.
Puromycin Dihydrochloride is a valuable tool for investigating protein synthesis, studying translation mechanisms, and selecting cells with specific genetic modifications.Specifications
Specifications
| Chemical Name or Material | Puromycin |
| CAS | 58-58-2 |
| Quantity | 100 mg |
| Molecular Formula | C22H29N7O5-2 HCl |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.