missing translation for 'onlineSavingsMsg'
Learn More

Puromycin aminonucleoside, MedChemExpress

Product Code. 16452827
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16452827

Brand: Medchem Express HY15695

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Puromycin aminonucleoside (NSC 3056) is the aminonucleoside portion of the antibiotic puromycin, and used in nephrosis animal models. Puromycin aminonucleoside induces apoptosis. Puromycin aminonucleoside is a reversible inhibitor of dipeptidyl peptidase II and cytosol alanyl aminopeptidase.

Puromycin aminonucleoside (NSC 3056) is the aminonucleoside portion of the antibiotic puromycin, and used in nephrosis animal models. Puromycin aminonucleoside induces apoptosis. Puromycin aminonucleoside is a reversible inhibitor of dipeptidyl peptidase II and cytosol alanyl aminopeptidase.

Specifications

Chemical Name or Material Puromycin aminonucleoside
Physical Form Solid
CAS 58-60-6
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C12H18N6O3
For Use With (Application) Cancer-Kinase/protease
Synonym NSC 3056
Solubility Information H2O : 40 mg/mL (135.91 mM; ultrasonic and warming and heat to 50°C) ∣DMSO : 25 mg/mL (84.94 mM; ultrasonic and warming and heat to 60°C)
SMILES OC[C@@H]1[C@@H](N)[C@H]([C@H](N2C=NC3=C2N=CN=C3N(C)C)O1)O
Molecular Weight (g/mol) 294.31
Formula Weight 294.31
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.