missing translation for 'onlineSavingsMsg'
Learn More

Euro Protection™ Purofort™ Safety Boots

Product Code. 15879226
Change view
Click to view available options
Size:
39
40
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15879226 40 Each
15869226 39 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15879226 Supplier Euro Protection™ Supplier No. 9PUSG40

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Euro Protection™ Purofort™ Safety Boots provide flexible and lightweight protection and cold insulation and feature a steel toe cap for protection.

  • Flexible and lightweight for use all day long
  • Cold insulation
  • Tear and wear resistant
  • Purofort™ upper
  • Polyamide lining
  • Polyurethane insole
  • Steel toe cap
  • Steel, puncture-resistant sole
  • Purofort™ outsole
  • Suitable for use in farming industry

Specifications

Color Green
For Use With (Application) Agriculture/Farming
Liner Material Polyamide
Size 40
Sole Features Polyurethane insole, steel, puncture-resistant sole, Purofort™ outsole.
Sole Material Purofort™
Toe Material Steel
Weight (Metric) 900 g per boot in size 9.5
Certifications/Compliance (Europe) EN ISO 20345 : 2011
Product Type Safety Boot
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.