missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Purmorphamine
GREENER_CHOICE

Product Code. 30002010
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30002010

Brand: Tocris Bioscience™ 4551/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Smo receptor agonist

Purmorphamine is a smoothened (Smo) receptor agonist (EC50 ∼ 1 μM) and activates the Hedgehog pathway. Purmorphamine induces osteogenesis in mouse mesenchymal progenitor cells (C3H10T1/2). When combined with BMP-4, Purmorphamine can transdifferentiate pre-adipocytes (3T3-L1) and myoblasts (C2C12) into osteoblasts. Purmorphamine induces differentiation of multipotent mesenchymal progenitor cells into osteoblasts, and of spinal motor neurons from pluripotent human stem cells, and protects dopaminergic neurons in a mouse model of Parkinson's disease.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 483367-10-8
Molecular Formula C31H32N6O2

Specifications

Quantity 10 mg
Formula Weight 520.62
Molecular Weight (g/mol) 520.637
Chemical Name or Material Purmorphamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.