missing translation for 'onlineSavingsMsg'
Learn More

Purmorphamine, MedChemExpress

Product Code. 16466318
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466318 10 mg 10mg
16422437 50 mg 50mg
16418907 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16466318 Supplier Medchem Express Supplier No. HY15108
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Purmorphamine is a smoothened/Smo receptor agonist with an EC50 of 1 μM.

Purmorphamine is a smoothened/Smo receptor agonist with an EC50 of 1 μM.

Specifications

Chemical Name or Material Purmorphamine
CAS 483367-10-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C31H32N6O2
Quantity 10 mg
Solubility Information DMSO : 33.33 mg/mL (64.02 mM; Need ultrasonic)
SMILES N1(C2=CC=C(NC3=NC(OC4=CC=CC5=C4C=CC=C5)=NC6=C3N=CN6C7CCCCC7)C=C2)CCOCC1
Molecular Weight (g/mol) 520.62
Formula Weight 520.62
Percent Purity 99.2%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.