missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Purmorphamine
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
- Purmorphamine is a tri-substituted purine derivative that activates the Hedgehog pathway by directly binding to and activating the Hedgehog receptor Smoothened (EC50 = 1 μM)
- (Sinha and Chen)
- Promotes differentiation of ventral spinal progenitors and motor neurons from human pluripotent stem cells (Hu and Zhang, Karumbayaram et al.), Li et al.)
- Promotes differentiation of osteoblasts from human and mouse mesenchymal cells (Beloti et al.), Wu et al. 2002, Wu et al.) 2004)
- Inhibits differentiation and maturation of adipocytes from human mesenchymal cells (Fontaine et al.)
Specifications
Specifications
| CAS | 483367-10-8 |
| Quantity | 1 mg |
| Molecular Formula | C31H32N6O2 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction