missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PURL Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94134-0.1ml
This item is not returnable.
View return policy
Description
PURL Polyclonal antibody specifically detects PURL in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PURL | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FGAM synthase, FGAMS, FGAR amidotransferase, formylglycinamide ribotide synthetase, KIAA0361, phosphoribosylformylglycinamidine synthase, PURL | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human PFAS (NP_036525.1). MSPVLHFYVRPSGHEGAAPGHTRRKLQGKLPELQGVETELCYNVNWTAEALPSAEETKKLMWLFGCPLLLDDVARESWLLPGSNDLLLEVGPRLNFSTPTSTNIVSVCRATGLGPVDRVETTRRYRLSFAHPPSAEVEAIALATLHDRMTEQHFPHPIQSFSPESMPEPLNGPINILGEGRLALEKANQELGLALDSWDLDFYTKRFQELQRNPSTVEAFDLAQSNSEHSRHWFFKGQLHVDGQKLVHSLFESIMSTQESSNPNNVLKFCDNSSAIQGKEVRFLRPEDPTRPSRFQQQQG | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
| 5198 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction