missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PURG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599039
This item is not returnable.
View return policy
Description
PURG Polyclonal Antibody for Western Blot, ELISA
The exact function of this gene is not known, however, its encoded product is highly similar to purine-rich element binding protein A. The latter is a DNA-binding protein which binds preferentially to the single strand of the purine-rich element termed PUR, and has been implicated in the control of both DNA replication and transcription. PURG lies in close proximity to the Werner syndrome gene, but on the opposite strand, on chromosome 8p11. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| PURG | |
| Polyclonal | |
| Unconjugated | |
| PURG | |
| 4930486B15Rik; MGC119274; Purg; PURG-A; Pur-gamma; PURG-B; purine rich element binding protein G; purine-rich element binding protein G; Purine-rich element-binding protein G; Purine-rich element-binding protein gamma | |
| Rabbit | |
| Protein G | |
| RUO | |
| 29942 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 4.92 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9UJV8 | |
| PURG | |
| Recombinant Human Purine-rich element-binding protein gamma protein (242-345AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction