missing translation for 'onlineSavingsMsg'
Learn More

HALYARD™ PUREZERO™ HG3 Light Blue Sterile Nitrile Gloves
GREENER_CHOICEBIOPHARMA

Product Code. 17364350 Shop All Halyard Products
Click to view available options
Size:
6
6.5
7
7.5
8
8.5
9
10
Unit Size:
300 pairs

Product Code. 17364350

Brand: HALYARD™ CLN923290

Only null left
Add to basket
Add to basket

High-quality gloves designed to the exact standards to meet the requirements of a contamination controlled environment.

A cleanroom glove with guaranteed low particulate counts and endotoxin levels due to repeated washes in deionized water and chlorination.

  • Nickel-free
  • Powder-free
  • Silicone-free
  • Latex-free and no Phtalates
  • High cleanliness
  • Low endotoxin levels
  • Formulated with ZERO presence of these hazardous traditional accelerators: Sulfur, Thiurams, Thiazoles, Guanidine, Carbamates(ZDEC or ZDBC)
  • Backed up by a vertically integrated supply chain

Specifications

Certifications/Compliance ISO 3, ISO 4, ISO 5, ISO 6, ISO 7
Certifications/Compliance (Europe) GMP Grade A/B/C/D
Chemotherapeutic Rated ASTM D6978
Chlorinated Yes
Cleanroom Class Class 10 (ISO 4)
Accelerator Free Yes
AQL 1
Color Light Blue
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 4 mil
Finger Thickness (Metric) 0.10 mm
Hand Specific
Latex Free Yes
Material Nitrile
Palm Thickness (English) 3.1 mil
Palm Thickness (Metric) 0.08 mm
Powdered Powder Free
Product Type Controlled Environment Gloves
Size 9
Sterility Sterile
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
Manufacturing/QMS Audit Standards ISO 9001:2015, EN13485:2016
Model CLN923290
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.