missing translation for 'onlineSavingsMsg'
Learn More

HALYARD™ PUREZERO™ HG3 Light Blue Sterile Nitrile Gloves
GREENER_CHOICEBIOPHARMA

Product Code. 17324350 Shop All Halyard Products
Change view
Click to view available options
Size:
6
6.5
7
7.5
8
8.5
9
10
Unit Size:
300 pairs
Product Code. Size unitSize
17324350 7 300 pairs
17304350 6 300 pairs
17314350 6.5 300 pairs
17334350 7.5 300 pairs
17344350 8 300 pairs
17354350 8.5 300 pairs
17364350 9 300 pairs
17394340 10 300 pairs
8 options

Product Code. 17324350

Brand: HALYARD™ CLN923270

Only null left
Add to basket
Add to basket

High-quality gloves designed to the exact standards to meet the requirements of a contamination controlled environment.

A cleanroom glove with guaranteed low particulate counts and endotoxin levels due to repeated washes in deionized water and chlorination.

  • Nickel-free
  • Powder-free
  • Silicone-free
  • Latex-free and no Phtalates
  • High cleanliness
  • Low endotoxin levels
  • Formulated with ZERO presence of these hazardous traditional accelerators: Sulfur, Thiurams, Thiazoles, Guanidine, Carbamates(ZDEC or ZDBC)
  • Backed up by a vertically integrated supply chain

Specifications

Certifications/Compliance ISO 3, ISO 4, ISO 5, ISO 6, ISO 7
Certifications/Compliance (Europe) GMP Grade A/B/C/D
Chemotherapeutic Rated ASTM D6978
Chlorinated Yes
Cleanroom Class Class 10 (ISO 4)
Accelerator Free Yes
AQL 1
Color Light Blue
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 4 mil
Finger Thickness (Metric) 0.10 mm
Hand Specific
Latex Free Yes
Material Nitrile
Palm Thickness (English) 3.1 mil
Palm Thickness (Metric) 0.08 mm
Powdered Powder Free
Product Type Controlled Environment Gloves
Size 7
Sterility Sterile
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
Manufacturing/QMS Audit Standards ISO 9001:2015, EN13485:2016
Model CLN923270
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.