Learn More
Applied Biosystems™ PureQuant CD4+ T Cell Assay
Shop All Applied Biosystems ProductsDescription
The Applied Biosystems PureQuant assays are a family of assays that accurately identify and quantify specific immune cell types in a mixed population of cells. These assays are TaqMan qPCR-based and measure genomic DNA methylation at specific sites that serve as a unique identifier. The assays include standards and controls and each 96-well plate can accommodate up to seven test samples. Results are provided as a percentage of target cell type out of total input cells using a free analysis tool.
The PureQuant CD4+ T-Cell Assay utilizes the CD4 gene as an epigenetic marker for CD4+ T cells; the assay targets regulatory elements within its first intron.
Benefits include:
- Proven performance―core technology has been used for identification and quantification of immune cell types for over a decade
- Easily standardized―qPCR method enables consistent results across multiple users, instruments, and sites
- Facilitates regulatory filings―regulatory support file provided upon request; established performance specifications following ICHQ2(R1) guidelines
- Scheduling flexibility―assay can be performed using fresh or frozen cells/gDNA
The assays include all reagents, standards, and controls required to run the assay. Specific components include:
- Calibrator: corrects for variation in bisulfite conversion efficiency. Plasmid-based calibrator harbors the genomic target regions in equimolar ratios and includes GAPDH. Subjected to bisulfite conversion followed by copy number estimation via qPCR standard curve. Allows estimation of assay-specific bisulfite efficiency factor: the ratio between TpG/GAPDH copy numbers measured from the same plasmid.
- Reference: checks assay performance. Consists of genomic DNA extracted from human blood of pooled healthy donors. The reference values represent the expected range of the target cell type. For example, CD8+ cells are generally more abundant than Treg or Th17 cells.
- Standards: determine the unknown copy number. Consist of six dilutions spanning 4-log linear dynamic range (30-31250 copies). Absolute copy numbers can reliably be determined using the slope and intercept of the respective standard curves.
This product is manufactured at a site with an ISO9001-certified quality management system.
Specifications
Specifications
| Content And Storage | • Dynabeads SILANE Genomic DNA Kit, store at 2–8°C • 1 x 2 mL PureQuant qPCR Master Mix, store at 2–8°C • 4 x ABS • THFA, store at room temperature • PureQuant CD4 Module, store at -15 to -25°C (CD4 and GAPDH TaqMan primers and probes, Reference DNA, Lambda DNA, Standard DNA) |
| Detection Method | Primer-probe |
| Form | Frozen |
| Format | 96-Well Plate |
| PCR Method | qPCR |
| Reaction Speed | Fast or Standard |
| For Use With (Equipment) | QuantStudio™ |
| Product Type | Purequant CD4 + T Cell Assay |
| Quantity | 28 reactions |
| No. of Reactions | 28 Reactions |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.