missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ PurePlus™ Solid Frame Real Time PCR Plates with Full Skirt
GREENER_CHOICE

Product Code. 30280311
Click to view available options
Array:
12 x 8
16 x 24
No. of Wells:
96
384
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 30280311

Brand: Labcon™ 39665200009

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Labcon consumables for qPCR are optimized for use in thermo cycling processing that use fluorescent dye or DNA probes.

  • 96 or 384 well skirted PCR plates for real-time PCR thermocyclers
  • Solid clear frame; White opaque wells
  • With reference grid
  • ATP, RNase/DNase, DNA and Endotoxin Free (non-pyrogenic)
  • Polypropylene USP Class VI resin
  • Freezable to -80°C
  • Made in USA

Specifications

Barcode No Barcode
Certifications/Compliance ISO 9001:2015, FDA CFR 21 GMP
Format SBS Standard
Array 16 x 24
Autoclavable Not Autoclavable
Packaging 10 Plates/Pk., 10 Pk./Cs.
Color Clear
Disposable Single-use
Product Type qPCR plate
Skirt Style Full skirt
Material Polycarbonate
Volume (Metric) Well 25 μL
No. of Wells 384
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) qPCR
High-throughput Compatibility Yes
RNase Free Yes
Well Color White
Fit for Purpose PCR Ready
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.