Learn More
Labcon™ PurePlus™ 1.2 mL Sample Library Tubes
Product Code. 17527936
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Individual Tubes, Rack
Strips of 12, Bagged
Strips of 12, Rack
Strips of 8, Rack
Unit Size:
Pack of 80
Pack of 960
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Format unitSize
17527936 Sterile Individual Tubes, Rack Pack of 960
17517936 Non-sterile Individual Tubes, Rack Pack of 960
17547936 Sterile Strips of 8, Rack Pack of 960
17537936 Non-sterile Strips of 8, Rack Pack of 960
17567936 Sterile Strips of 12, Rack Pack of 960
17557936 Non-sterile Strips of 12, Rack Pack of 960
17517946 Non-sterile Strips of 12, Bagged Pack of 80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 17527936 Supplier Labcon™ Supplier No. 39105450009
Only null left
Add to basket
Add to basket
Sample library tubes come in strips of 8, 12, or as individual tubes.
  • Available in 96-place racks or resealable bags
  • ATP, RNase/DNase, Endotoxin-free (non-pyrogenic)
Specifications
Autoclavable Autoclavable
Color Clear
Material Polypropylene
Format Individual Tubes, Rack
No. per Pack 960/Pk.
Packaging 960 Tubes/Pk., 10 Pk./Cs.
Volume (Metric) 1.2 mL
Sterility Sterile