missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PureLink™ Fast Low-Endotoxin Midi Plasmid Purification Kit
The PureLink Fast Low-Endotoxin Midi Plasmid Purification Kit uses next-generation columns with advanced silica membranes that enable isolation of transfection grade (<1 EU/μg) plasmid DNA in about 30 minutes.
£236.00 - £432.00
Specifications
Content And Storage | • Resuspension buffer, store at 4°C upon reciept • Lysis buffer, store at room temperature • Precipitation buffer, store at room temperature • Binding buffer, store at room temperature • Wash buffer 1, store at room temperature • Wash buffer 2 (concentrate), store at room temperature • Elution buffer, store at room temperature • Column assemblies, store at room temperature • Syringe filters, store at room temperature • Syringe plungers, store at room temperature • Collection tubes, store at room temperature |
---|---|
Format | Kit |
Isolation Technology | Silica Column |
Scale | Midi |
For Use With (Application) | Cloning, PCR |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
15900493
|
Invitrogen™
A35892 |
25 Preps |
£236.00
Pack of 25 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17113129
|
Invitrogen™
A36227 |
50 Preps |
£432.00
Pack of 50 |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The simple precipitation-free protocol yields up to 0.4 mg plasmid, suitable for standard transfections and all molecular biology applications such as cloning and sequencing.
- Ultra-fast protocol—innovative silica-based columns enable purification of transfection grade plasmid DNA in under 30 minutes
- Low endotoxin plasmid—the resulting plasmid contains less than 1 EU/μg endotoxins
- High yields—isolate up to 0.4 mg of high-quality plasmid DNA
- Colored buffers—the kit features colored buffers that permit error-free visualization of complete bacterial cell lysis and subsequent neutralization
Specifications
• Resuspension buffer, store at 4°C upon reciept • Lysis buffer, store at room temperature • Precipitation buffer, store at room temperature • Binding buffer, store at room temperature • Wash buffer 1, store at room temperature • Wash buffer 2 (concentrate), store at room temperature • Elution buffer, store at room temperature • Column assemblies, store at room temperature • Syringe filters, store at room temperature • Syringe plungers, store at room temperature • Collection tubes, store at room temperature |
|
Silica Column | |
Cloning, PCR | |
PureLink™ | |
Room Temperature |
Kit | |
Midi | |
Midi scale input culture of 25-100 mL to obtain yields of up to 200 μg | |
Plasmid Purification Kit | |
30 min. |
For Research Use Only. Not for use in diagnostic procedures.