missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Fast Low-Endotoxin Maxi Plasmid Purification Kit

Product Code. 15974140
Change view
Click to view available options
No. of Reactions:
10 Preps
20 Preps
Unit Size:
Pack of 10
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
15974140 20 Preps Pack of 20
15801782 10 Preps Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15974140 Supplier Invitrogen™ Supplier No. A36228

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PureLink Fast Low-Endotoxin Maxi Plasmid Purification Kit uses next-generation columns with advanced silica membranes that enable isolation of transfection grade (<1 EU/μg) plasmid DNA in about 30 minutes.

The simple precipitation-free protocol yields up to 1.5 mg plasmid, suitable for standard transfections and all molecular biology applications such as cloning and sequencing.

  • Ultra-fast protocol—innovative silica-based columns enable purification of transfection grade plasmid DNA in under 30 minutes
  • Low endotoxin plasmid—the resulting plasmid contains less than 1 EU/μg endotoxins
  • High yields—isolate up to 1.5 mg of high-quality plasmid DNA
  • Colored buffers—the kit features colored buffers that permit error-free visualization of complete bacterial cell lysis and subsequent neutralization
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Resuspension buffer, store at 4°C upon reciept
• Lysis buffer, store at room temperature
• Precipitation buffer, store at room temperature
• Binding buffer, store at room temperature
• Wash buffer 1, store at room temperature
• Wash buffer 2 (concentrate), store at room temperature
• Elution buffer, store at room temperature
• Column assemblies, store at room temperature
• Syringe filters, store at room temperature
• Syringe plungers, store at room temperature
• Collection tubes, store at room temperature

Format Kit
Isolation Technology Silica Column
Scale Maxi
For Use With (Application) Cloning, PCR
No. of Reactions 20 Preps
Prep Scale Maxi input culture of 100-500 mL to obtain a yield of up to 850 μg
Product Line PureLink
Product Type Plasmid Purification Kit
Quantity 20 Preps
Shipping Condition Room Temperature
Test Time 30 min.
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • X20 PL Fast Low-Endotoxin Maxi Plasmid Purificatio n Kit
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide, proteinase

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.