missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beskrivelse
Cadherin-11 Monoclonal antibody specifically detects Cadherin-11 in Human, Rat samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Tekniske data
Tekniske data
| Antigen | Cadherin-11 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | 16A |
| Conjugate | FITC |
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen |
| Formulation | PBS |
| Gene Alias | CAD11OSF-4, cadherin 11, type 2, OB-cadherin (osteoblast), cadherin-11, CDHOB, OB, OB-cadherin, Osteoblast cadherin |
| Host Species | Mouse |
| Immunogen | This antibody is a mouse monoclonal IgG1 antibody obtained by fusion of SP2/0 Mouse myeloma cells with spleen cells from a BALB/c Mouse immunized with affinity purified extracellular domain of Human cadherin-11-GST fusion protein. |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?