missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PureCoat™ 384-Well, Amine-Treated, Flat-Bottom Microplate
Click to view available options
Packaging:
5/Cs
50/Cs
Description
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
Specifications
Format | Standard |
Lid | With Lid |
Array | 16 x 24 |
Packaging | 5/Cs |
Color | Black, Clear Bottom |
Material | Polystyrene |
No. of Wells | 384 |
Sterility | Non-sterile |
Surface Treatment | Amine |
Well Shape | Flat Bottom (F-well) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'