Learn More
Saint Gobain Life Sciences Pure-Fit SIB Polypropylene
Description
Pure-Fit™ TC tube clamps represent a new generation of clamping technology for the pharmaceutical industry. They feature an outer body with no sharp edges, eliminating the potential for snags, scratches, or punctures; a press-down locking system that is fully encapsulated to guard against untimely or unwanted release; a side release mechanism that facilitates secure, single-handed installation and operation; and an integral bore hole for absolute lock-out capabilities.
- Provides complete fluid stoppage
- Smooth edges and corners help prevent punctures and ruptures
- Can be installed on or over tubing or finished assemblies
- High degree of leverage accommodates tubing with different durometers
- Single-hand installation capability for ease of operation
- Unique top locking mechanism and side release prevent unwanted openings
- Available in PVDF or polypropylene
- Fully autoclavable and sterilizable
- Custom colors available
Applications
- Pharmaceutical Processes
- Peristaltic Pump Sets
- Drug Delivery and Discovery
- Medical Systems
- Laboratory Functions
- Assemblies and Tubing Sets
Specifications
Specifications
| Autoclavable | Autoclavable |
| Quantity | 25/Pk. |
| Type | Tubing Clamp |
| Material | Polypropylene |
| Color | Clear |
| Jaw Type | Standard Taper |
| Diameter (Metric) Inside | 3/8 to 3/4 in. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.