missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Pure 70mm Diameter Stainless Steel Ring Sieve For Ultra Centrifugal Mill ZM 200

Product Code. 11711856 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
0.12 mm
0.2 mm
0.25 mm
0.28 mm
0.5 mm
0.75 mm
1 mm
1.5 mm
2 mm
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size unitSize
11711856 0.5 mm Each
10732847 0.12 mm Each
11721856 0.25 mm Each
10401705 0.2 mm Each
10147164 0.28 mm Each
11701856 0.75 mm Each
11944272 1.5 mm Each
11964272 2 mm Each
10650974 1 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 11711856 Supplier RETSCH Supplier No. 03.647.0285
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Quantity 1Each
Pore Size 0.5 mm
Type Ring Sieve
Material Stainless Steel
For Use With (Equipment) Ultra Centrifugal Mill ZM 200
Diameter (Metric) Outer 70 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.