missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PURB Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 78%; Zebrafish: 100%.
This gene product is a sequence-specific, single-stranded DNA-binding protein. It binds preferentially to the single strand of the purine-rich element termed PUR, which is present at origins of replication and in gene flanking regions in a variety of eukaryotes from yeasts through humans. Thus, it is implicated in the control of both DNA replication and transcription. Deletion of this gene has been associated with myelodysplastic syndrome and acute myelogenous leukemia.
Specifications
Specifications
| Antigen | PURB |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | PURB |
| Gene Accession No. | Q96QR8 |
| Gene Alias | 2310015K15Rik; AA114818; Cager-2; D11Bwg0414e; LOW QUALITY PROTEIN: transcriptional activator protein Pur-beta; purb; PurB beta; purb protein; purbb; PURBETA; pur-beta; purine rich element binding protein B; purine-rich element binding protein B; purine-rich element binding protein Bb; purine-rich element-binding protein B; purine-rich single-stranded DNA-binding protein beta; transcription factor Pur-beta; transcriptional activator protein Pur-beta; Vascular actin single-stranded DNA-binding factor 2 p44 component; zgc:65916 |
| Gene Symbols | PURB |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?