missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PURB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-99780
This item is not returnable.
View return policy
Description
PURB Polyclonal specifically detects PURB in Human samples. It is validated for Immunoprecipitation.
Specifications
| PURB | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| PURB | |
| The immunogen this antibody was made to, region between residue 1 and 50 of human Purine-rich Element Binding Protein B using the numbering given in entry NP_150093.1 (GeneID 5814). | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 5814 | |
| Human | |
| IgG |
| Immunoprecipitation | |
| 1.0 mg/mL | |
| Immunoprecipitation 2-10 mcg/mg lysate | |
| MGC126784, PURBETA, purine-rich element binding protein B | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| NP_150093.1. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction