missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PURA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17733-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene product is a sequence-specific, single-stranded DNA-binding protein. It binds preferentially to the single strand of the purine-rich element termed PUR, which is present at origins of replication and in gene flanking regions in a variety of eukaryotes from yeasts through humans. Thus, it is implicated in the control of both DNA replication and transcription. Deletion of this gene has been associated with myelodysplastic syndrome and acute myelogenous leukemia.Specifications
| PURA | |
| Polyclonal | |
| Unconjugated | |
| PURA | |
| PUR ALPHA, PUR1, PURA, PURALPHA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19290, 307498, 5813 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P42669, P86252, Q00577 | |
| PURA | |
| PURA Fusion Protein Ag12173 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction