missing translation for 'onlineSavingsMsg'
Learn More

BM Polyco™ Pura™ PVC Flock-Lined Gloves

Product Code. 12930775
Change view
Click to view available options
Size:
7
8
9
10
Unit Size:
12 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12930775 7 12 pairs
12940775 8 12 pairs
12950775 9 12 pairs
12960775 10 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12930775 Supplier BM Polyco™ Supplier No. 474
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

BM Polyco™ Pura™ PVC Flock-Lined Gloves are medium-weight gloves ideal for general cleaning work or food handling

  • PVC gloves provide an excellent alternative for those allergic to natural latex proteins or sensitive to the chemical accelerators used in other types of rubber gloves
  • Anatomically shaped for additional comfort
  • Slip resistant pattern on palm enhances grip
  • Soft flock lining absorbs perspiration keeping hands cool and comfortable
  • Tested in accordance with EN1186, making them suitable for food contact
  • Also ideal for general cleaning and janitorial duties
  • Available in a wide range of colours for easy job application segregation

Specifications

Certifications/Compliance CE, EN1186, EN388, EN374-2, EN374-3
Color Blue
Disposable Reusable
Hand Ambidextrous
Latex Free Yes
Material PVC
Powdered Powder Free
Size 7
Sterility Non-sterile
For Use With (Application) General Purpose Cleaning Glove
Glove Cuff Style Plain
Liner Material Flock
Lining Flock Lined
Quantity 12/Pk.
Length (Metric) 310 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.