Learn More
Invitrogen™ PURA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113567
Description
Immunogen sequence: VEFRDYLGDF IEHYAQLGPS QPPDLAQAQD EPRRALKSEF LVRENRKYYM For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%.
This gene product is a sequence-specific, single-stranded DNA-binding protein. It binds preferentially to the single strand of the purine-rich element termed PUR, which is present at origins of replication and in gene flanking regions in a variety of eukaryotes from yeasts through humans. Thus, it is implicated in the control of both DNA replication and transcription. Deletion of this gene has been associated with myelodysplastic syndrome and acute myelogenous leukemia.
Specifications
| PURA | |
| Polyclonal | |
| Unconjugated | |
| PURA | |
| CAGER-1; MRD31; Pur alpha; PUR1; Pura; pura {ECO:0000250; PURALPHA; Pur-alpha; purine rich element binding protein A; purine-rich element binding protein A; purine-rich single-stranded DNA-binding protein alpha; purine-rich single-stranded DNA-binding protein alpha {ECO:0000250; ssCRE-BP; transcriptional activator protein Pur-alpha; transcriptional activator protein Pur-alpha {ECO:0000250; UniProtKB:P42669} | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5813 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q00577 | |
| PURA | |
| Synthetic peptide directed towards the middle region of human PURA. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.